missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydronaphthalene 98.0+%, TCI America™

Catalog No. T0713250ML
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0713250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T0713250ML Supplier TCI America Supplier No. T0713250ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 119-64-2
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00001733
InChI Key CXWXQJXEFPUFDZ-UHFFFAOYSA-N
Synonym tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene
PubChem CID 8404
ChEBI CHEBI:35008
IUPAC Name 1,2,3,4-tetrahydronaphthalene
SMILES C1CCC2=CC=CC=C2C1

Specifications

Color Colorless
Boiling Point 207°C
Quantity 250 mL
UN Number 3082
Formula Weight 132.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3,4-Tetrahydronaphthalene [for Spectrophotometry]
EINECSNumber (4)-0574
RTECSNumber QK3850000
TSCA Yes