missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,5,6-Hexa-O-[11-[4-(4-hexylphenylazo)phenoxy]undecanoyl]-D-mannitol 97.0+%, TCI America™

Catalog No. H14525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14525G 5 g
H14521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14525G Supplier TCI America Supplier No. H14525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1093077-77-0
Molecular Formula C180H254N12O18
Molecular Weight (g/mol) 2874.078
InChI Key DMCHOYXACRXKRI-RDXCSIDZSA-N
PubChem CID 91972138
SMILES CCCCCCC1=CC=C(C=C1)N=NC2=CC=C(C=C2)OCCCCCCCCCCC(=O)OCC(C(C(C(COC(=O)CCCCCCCCCCOC3=CC=C(C=C3)N=NC4=CC=C(C=C4)CCCCCC)OC(=O)CCCCCCCCCCOC5=CC=C(C=C5)N=NC6=CC=C(C=C6)CCCCCC)OC(=O)CCCCCCCCCCOC7=CC=C(C=C7)N=NC8=CC=C(C=C8)CCCCCC)OC(=O)CCCCCCCCCCOC9=CC=C(C=C9)N=NC

Specifications

Quantity 5 g
Formula Weight 2874.08
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4,5,6-Hexa-O-[11-[4-(4-hexylphenylazo)phenoxy]undecanoyl]-D-mannitol