missing translation for 'onlineSavingsMsg'
Learn More

1,2,2,3-Tetrachloropropane 95.0+%, TCI America™

Catalog No. T070810ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T070810ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T070810ML Supplier TCI America Supplier No. T070810ML

Chemical Identifiers

CAS 13116-53-5
Molecular Formula C3H4Cl4
Molecular Weight (g/mol) 181.865
MDL Number MFCD00039362
InChI Key UDPHJTAYHSSOQB-UHFFFAOYSA-N
PubChem CID 25717
IUPAC Name 1,2,2,3-tetrachloropropane
SMILES C(C(CCl)(Cl)Cl)Cl

Specifications

Color Yellow
Boiling Point 164°C
Quantity 10 mL
Formula Weight 181.87
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,2,3-Tetrachloropropane
EINECSNumber (2)-0083
RTECSNumber TZ7000100
TSCA Yes