missing translation for 'onlineSavingsMsg'
Learn More

1,11-Undecanedicarboxylic acid, Thermo Scientific™

Catalog No. p-2689402
Change view
Click to view available options
Quantité:
5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
AC369440050 5g
AC369440250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC369440050 Supplier Thermo Scientific Chemicals Supplier No. 369440050

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 505-52-2
Formule moléculaire C13H24O4
Poids moléculaire (g/mol) 244.33
Numéro MDL MFCD00002740
Clé InChI DXNCZXXFRKPEPY-UHFFFAOYSA-N
Synonyme brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
PubChem CID 10458
ChEBI CHEBI:73718
Nom de l’IUPAC tridecanedioic acid
SOURIRES C(CCCCCC(=O)O)CCCCCC(=O)O

Specifications

Nom ou substance chimique 1, 11-Undecanedicarboxylic acid
Nom Remarque 94%
CAS % minimum 92.0
CAS % maximal 100.0
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 94%
Emballage Glass bottle
Formule linéaire HO2C(CH2)11CO2H
Quantité 5g
Beilstein 02,731
Renseignements sur la solubilité Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether
Poids de formule 244.33
Pourcentage de pureté ≥92%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/pro
EINECSNumber 208-011-4

RUO – Research Use Only