missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I) 95.0+%, TCI America™
Supplier: TCI America P21721G
Specifications
| (1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I) | |
| C31H23CuF3N2P | |
| 1 g | |
| [Cu+].F[C-](F)F.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 575.05 | |
| 575.05 | |
| Crystalline Powder |
| 1334890-93-5 | |
| MFCD28386111 | |
| AKGOGTMOALVICI-UHFFFAOYSA-N | |
| λ¹-copper(1+) 1,10-phenanthroline trifluoromethanide triphenylphosphane | |
| 88921701 | |
| ≥95.0% (T) |
Chemical Identifiers
| 1334890-93-5 | |
| 575.05 | |
| AKGOGTMOALVICI-UHFFFAOYSA-N | |
| λ¹-copper(1+) 1,10-phenanthroline trifluoromethanide triphenylphosphane |
| C31H23CuF3N2P | |
| MFCD28386111 | |
| 88921701 | |
| [Cu+].F[C-](F)F.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
TSCA : No