missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I) 95.0+%, TCI America™
$746.97 - $2589.36
Chemical Identifiers
| CAS | 1334890-93-5 |
|---|---|
| Molecular Formula | C31H23CuF3N2P |
| Molecular Weight (g/mol) | 575.05 |
| MDL Number | MFCD28386111 |
| InChI Key | AKGOGTMOALVICI-UHFFFAOYSA-N |
| PubChem CID | 88921701 |
| IUPAC Name | λ¹-copper(1+) 1,10-phenanthroline trifluoromethanide triphenylphosphane |
| SMILES | [Cu+].F[C-](F)F.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 1334890-93-5 | |
| 575.05 | |
| AKGOGTMOALVICI-UHFFFAOYSA-N | |
| λ¹-copper(1+) 1,10-phenanthroline trifluoromethanide triphenylphosphane |
| C31H23CuF3N2P | |
| MFCD28386111 | |
| 88921701 | |
| [Cu+].F[C-](F)F.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
| 1334890-93-5 | |
| MFCD28386111 | |
| AKGOGTMOALVICI-UHFFFAOYSA-N | |
| λ¹-copper(1+) 1,10-phenanthroline trifluoromethanide triphenylphosphane | |
| 88921701 | |
| ≥95.0% (T) | |
| (1,10-Phenanthroline)(trifluoromethyl)(triphenylphosphine)copper(I) |
| C31H23CuF3N2P | |
| 1 g | |
| [Cu+].F[C-](F)F.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 575.05 | |
| 575.05 | |
| Crystalline Powder |
Safety and Handling
TSCA : No