accessibility menu, dialog, popup

UserName

CAS RN 1334890-93-5

PNNCFFFCu

CAS RN 1334890-93-5

IUPAC Name: λ¹-copper(1+) 1,10-phenanthroline trifluoromethanide triphenylphosphane
Synonyms: λ¹-copper(1+) phen trifluoromethanide triphenylphosphine
Molecular Weight (g/mol): 575.05
Molecular Formula: C31H23CuF3N2P
InChi Key: AKGOGTMOALVICI-UHFFFAOYSA-N
SMILES: [Cu+].F[C-](F)F.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results