missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline monohydrate, For the Spectrophotometric Determination of Fe, Pd, V, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111020158
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020158 5 g
111018525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020158 Supplier Merck Emd Millipore Supplier No. 775005GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

1,10-Phenanthroline monohydrate is a chelating agent.

Chemical Identifiers

MDL Number MFCD00149973
Synonym o-Phenanthroline monohydrate

Specifications

Melting Point 100°C to 104°C; 99°C to 104°C
CAS 5144-89-8
Linear Formula C12H8N2 · H2O
Quantity 5 g
Formula Weight 198.22
Percent Purity ≥99.0% (HPLC); ≥99.0% (NT); ≥99.0%
RTECSNumber SF8437000