missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline Hydrochloride Monohydrate 99.0+%, TCI America™

Catalog No. P008125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P008125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P008125G Supplier TCI America Supplier No. P008125G
Only null left

Chemical Identifiers

CAS 3829-86-5
Molecular Formula C12H9ClN2
Molecular Weight (g/mol) 216.668
MDL Number MFCD00150061
InChI Key QPXDKQBBJCTNOY-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrochloride, 1,10-phenanthroline, monohydrochloride, phenanthroline hydrochloride, phen hydrochloride, 1,10-diazaphenanthrene hydrochloride, 1,10-phenanthroline, hydrochloride 1:1, 66-71-7 parent, o-phenanthrolinehydrochloride, c12h8n2.hcl, 1,10-phenanthrolinium chloride
PubChem CID 73350
IUPAC Name 1,10-phenanthroline;hydrochloride
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.Cl

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 216.67
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,10-Phenanthroline Hydrochloride Monohydrate