missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline, 99+%

Catalog No. AC157535000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC157535000 500 g
AC157530050 5 g
AC157530250 25 g
AC157531000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC157535000 Supplier Thermo Scientific Chemicals Supplier No. 157535000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 66-71-7 | C12H8N2 | 180.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66-71-7
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00011678
InChI Key DGEZNRSVGBDHLK-UHFFFAOYSA-N
Synonym o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, anhydrous, .beta.-phenanthroline
PubChem CID 1318
ChEBI CHEBI:44975
IUPAC Name 1,10-phenanthroline
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 117.0°C to 120.0°C
Color Pink or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 500 g
Beilstein 23,227
Merck Index 15,7328
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, acetone, ether and benzene
Formula Weight 180.21
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 1, 10-Phenanthroline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-629-2
RTECSNumber SF8300000
TSCA TSCA

RUO – Research Use Only