missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline, 0.1% (w/v), Ricca Chemical

Catalog No. p-3251189
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
55204 120 mL Natural Poly Bottle
552016 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55204 Supplier Ricca Chemical Company Supplier No. 55204
Only null left

Chemical Identifiers

CAS 5144-89-8 , 7732-18-5 , 7647-01-0
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
PubChem CID 21226
IUPAC Name 1,10-phenanthroline hydrate
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Color Colorless to Green
CAS Min % 0.1, 98.0, 0.12
CAS Max % 0.1, 100.0, 0.12
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 0.1% (w/v)
Chemical Name or Material 1, 10-Phenanthroline
Show More Show Less