missing translation for 'onlineSavingsMsg'
Learn More

1,10-Dichlorodecane 97.0+%, TCI America™

Catalog No. D0247500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0247500G 500 g
D024725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0247500G Supplier TCI America Supplier No. D0247500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2162-98-3
Molecular Formula C10H20Cl2
Molecular Weight (g/mol) 211.17
MDL Number MFCD00000957
InChI Key RBBNTRDPSVZESY-UHFFFAOYSA-N
PubChem CID 75101
IUPAC Name 1,10-dichlorodecane
SMILES ClCCCCCCCCCCCl

Specifications

Color Colorless
Boiling Point 102°C
Quantity 500 g
UN Number 3082
Synonym decane, 1,10-dichloro, decamethylene dichloride, decane,10-dichloro, 1, 10-dichlorodecane, acmc-1cbxv, ksc202q6p, 1,10-dichlorodecane, 1,10-decamethylene dichloride
Formula Weight 211.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,10-Dichlorodecane
EINECSNumber (2)-0066
TSCA Yes