missing translation for 'onlineSavingsMsg'
Learn More

1,10-Diaminodecane 98.0+%, TCI America™

Catalog No. D008525G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D008525G 25 g
D0085100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D008525G Supplier TCI America Supplier No. D008525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 646-25-3
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.316
MDL Number MFCD00008151
InChI Key YQLZOAVZWJBZSY-UHFFFAOYSA-N
Synonym 1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine
PubChem CID 1317
IUPAC Name decane-1,10-diamine
SMILES C(CCCCCN)CCCCN

Specifications

Melting Point 61°C
Color White-Yellow
Boiling Point 140°C
Quantity 25 g
UN Number 3259
Formula Weight 172.32
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,10-Diaminodecane
EINECSNumber (2)-3742
RTECSNumber HD7175000
TSCA Yes