missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenylethanol 98.0+%, TCI America™

Catalog No. D02955G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D02955G 5 g
D029525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D02955G Supplier TCI America Supplier No. D02955G

Chemical Identifiers

CAS 599-67-7
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.27
MDL Number MFCD00004450
InChI Key GIMDPFBLSKQRNP-UHFFFAOYSA-N
Synonym diphenylmethylcarbinol, methyldiphenylcarbinol, 1,1-diphenyl-1-ethanol, alpha-methylbenzhydrol, benzhydrol, .alpha.-methyl, benzenemethanol, .alpha.-methyl-.alpha.-phenyl, .alpha.-methylbenzhydrol, 1,1-diphenylethan-1-ol, unii-8v681bnm35, nsc33
PubChem CID 69031
IUPAC Name 1,1-diphenylethan-1-ol
SMILES CC(O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 82°C
Color White-Yellow
Boiling Point 155°C
Quantity 5 g
Formula Weight 198.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Diphenylethanol