missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenyl-2-picrylhydrazyl Free Radical 97.0+%, TCI America™

Catalog No. D43135G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43135G 5 g
D43131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43135G Supplier TCI America Supplier No. D43135G

1,1-Diphenyl-2-picrylhydrazyl Free Radical 97.0+%

Chemical Identifiers

CAS 1898-66-4
Molecular Formula C18H12N5O6
MDL Number MFCD00007231
Synonym DPPH

Specifications

Melting Point 139°C
Color Black
Quantity 5 g
Formula Weight 394.32
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Diphenyl-2-picrylhydrazyl Free Radical
RTECSNumber MW3250000
TSCA Yes
Recommended Storage Refrigerator