missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Diheptyl-4,4'-bipyridinium Dibromide 98.0+%, TCI America™

Catalog No. D15935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15935G 5 g
D159325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15935G Supplier TCI America Supplier No. D15935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6159-05-3 | C24H38Br2N2 | 514.39 g/mol

Chemical Identifiers

CAS 6159-05-3
Molecular Formula C24H38Br2N2
Molecular Weight (g/mol) 514.39
MDL Number MFCD00011997
InChI Key VRXAJMCFEOESJO-UHFFFAOYSA-L
Synonym 1,1'-diheptyl-4,4'-bipyridinium dibromide, dhbp dibromide, 1,1'-diheptyl-4,4'-bipyridine-1,1'-diium bromide, 4,4'-bipyridinium, 1,1'-diheptyl-, dibromide, 1,1'-di-n-heptyl-4,4'-bipyridinium dibromide, 4,4'-bipyridinium, 1,1'-diheptyl-, bromide 1:2, 1,1'-diheptyl-4,4'-bipyridine-1,1'-diium dibromide, 1-heptyl-4-1-heptylpyridin-1-ium-4-yl pyridin-1-ium dibromide, n,n'-diheptyl-4,4'-bipyridinium dibromide
PubChem CID 80262
IUPAC Name 1-heptyl-4-(1-heptylpyridin-1-ium-4-yl)pyridin-1-ium;dibromide
SMILES CCCCCCC[N+]1=CC=C(C=C1)C2=CC=[N+](C=C2)CCCCCCC.[Br-].[Br-]

Specifications

Quantity 5 g
Formula Weight 514.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Diheptyl-4,4′-bipyridinium Dibromide [for Electrochromic Material]