missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1'-Dibutyl-3,3,3',3'-tetramethylindotricarbocyanine Hexafluorophosphate 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 638.72 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,1′-Dibutyl-3,3,3′,3′-tetramethylindotricarbocyanine Hexafluorophosphate |
Safety and Handling
| TSCA | No |