missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Dibenzoylferrocene 98.0+%, TCI America™

Catalog No. B09155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09155G 5 g
B091525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09155G Supplier TCI America Supplier No. B09155G

Chemical Identifiers

CAS 12180-80-2
Molecular Formula C24H18FeO2
MDL Number MFCD00045455
Synonym Bis(benzoylcyclopentadienyl) Iron, Di(benzoylcyclopentadienyl) Iron

Specifications

Melting Point 103°C
Quantity 5 g
Formula Weight 394.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Dibenzoylferrocene