missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclohexanedicarboxylic acid, 95%

Catalog No. AAH2602503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2602503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2602503 Supplier Thermo Scientific Chemicals Supplier No. H2602503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1127-08-8 | C8H12O4 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1127-08-8
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00094668
InChI Key QYQADNCHXSEGJT-UHFFFAOYSA-N
Synonym cyclohexanedicarboxylic acid, 1,1-cyclohexanedicarboxylic acid, acmc-20ald2, cyclohexane dicarboxylic acid, 1,1-cyclohexanedicarboxylicacid, cyclohexane-1,1-dicarboxylicacid, cyclohexane-1,1-dicarboxylic acid
PubChem CID 136906
IUPAC Name cyclohexane-1,1-dicarboxylic acid
SMILES C1CCC(CC1)(C(=O)O)C(=O)O

Specifications

Melting Point ∼175°C (decomposition)
Odor Odorless
Quantity 1 g
Formula Weight 172.18
Percent Purity 95%
Chemical Name or Material 1,1-Cyclohexanedicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only