missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Benzothiophen-5-yl)-2-bromo-1-ethanone, 97%, Thermo Scientific™

Catalog No. CC24805CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC24805CB 250 mg Amber Glass Bottle
CC24805DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC24805CB Supplier Thermo Scientific Chemicals Supplier No. CC24805CB
Only null left

Chemical Identifiers

CAS 1131-87-9
Molecular Formula C10H7BrOS
Molecular Weight (g/mol) 255.13
MDL Number MFCD07368508
InChI Key NTQPGUCRHJHYIA-UHFFFAOYSA-N
Synonym 1-1-benzothiophen-5-yl-2-bromo-1-ethanone, 1-1-benzothiophen-5-yl-2-bromoethanone, 1-benzo b thiophen-5-yl-2-bromoethan-1-one, 5-bromoacetylbenzo b thiophene, 5-bromoacetyl-benzo b thiophene, 5-bromoacetyl benzo b thiophene, ethanone,1-benzo b thien-5-yl-2-bromo, 1-benzo b thiophen-5-yl-2-bromoethanone, 1-1-benzothiophen-5-yl-2-bromoethan-1-one
PubChem CID 7060546
IUPAC Name 1-(1-benzothiophen-5-yl)-2-bromoethanone
SMILES BrCC(=O)C1=CC=C2SC=CC2=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 255.13
Percent Purity 97%
Chemical Name or Material 1-(1-Benzothiophen-5-yl)-2-bromo-1-ethanone