missing translation for 'onlineSavingsMsg'
Learn More

1,1,3-Triphenyl-2-propyn-1-ol 98.0+%, TCI America™

Catalog No. T24185G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24185G Supplier TCI America Supplier No. T24185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 1522-13-0
Melting Point 82°C
Color White-Yellow
Boiling Point 220°C
Molecular Formula C21H16O
Quantity 5 g
MDL Number MFCD00004449
Synonym 1,1,3-triphenylpropargyl alcohol, 1,1,3-triphenyl-2-propyn-1-ol, diphenyl phenylethynyl carbinol, benzenemethanol, .alpha.-phenyl-.alpha.-phenylethynyl, triphenylprop-2-yn-1-ol, 1,1,3-triphenylpropargylalcohol, 2-propyn-1-ol, 1,1,3-triphenyl, pubchem14025, acmc-209d7i, triphenyl-1-propyne-3-ol
InChI Key VWRQCJRTHKUVNF-UHFFFAOYSA-N
SMILES OC(C#CC1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name triphenylprop-2-yn-1-ol
Molecular Weight (g/mol) 284.36
PubChem CID 137058
Formula Weight 284.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,3-Triphenyl-2-propyn-1-ol
Show More Show Less