missing translation for 'onlineSavingsMsg'
Learn More

1-(1,3-Benzothiazol-2-yl)-2-bromo-1-ethanone, 90+%, Thermo Scientific™

Catalog No. p-4509932
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC06005DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC06005DA Supplier Thermo Scientific Chemicals Supplier No. CC06005DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54223-20-0
Molecular Formula C9H6BrNOS
Molecular Weight (g/mol) 256.12
MDL Number MFCD03659698
InChI Key AYWGYNKWZWBMSV-UHFFFAOYSA-N
Synonym 1-1,3-benzothiazol-2-yl-2-bromo-1-ethanone, 1-benzo d thiazol-2-yl-2-bromoethanone, 1-1,3-benzothiazol-2-yl-2-bromoethanone, 2-bromoacetyl benzothiazole, 1-benzothiazol-2-yl-2-bromoethanone, 1-benzothiazol-2-yl-2-bromo-ethanone, 1-1,3-benzothiazol-2-yl-2-bromoethan-1-one, pubchem23327, 2-bromoacetyl-1,3-benzothiazole
PubChem CID 2776256
SMILES BrCC(=O)C1=NC2=CC=CC=C2S1

Specifications

Assay Percent Range 90%
Quantity 1 g
IUPAC Name 1-(1,3-benzothiazol-2-yl)-2-bromoethanone
Formula Weight 256.12
Percent Purity 90%
Chemical Name or Material 1-(1, 3-Benzothiazol-2-yl)-2-bromo-1-ethanone