missing translation for 'onlineSavingsMsg'
Learn More

1-(1,3-Benzodioxol-5-yl)-2-bromoethan-1-one, Thermo Scientific™

Catalog No. CC01605CB
Change view
Click to view available options
Quantity:
250mg
1g
10g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC01605CB 250mg Amber Glass Bottle
CC01605DA 1g Amber Glass Bottle
CC01605EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC01605CB Supplier Thermo Scientific Chemicals Supplier No. CC01605CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40288-65-1
Molecular Formula C9H7BrO3
Molecular Weight (g/mol) 243.056
InChI Key QBXCVQVFPVXAGS-UHFFFAOYSA-N
Synonym 1-1,3-benzodioxol-5-yl-2-bromoethan-1-one, 1-1,3-benzodioxol-5-yl-2-bromoethanone, 1-2h-1,3-benzodioxol-5-yl-2-bromoethan-1-one, 5-bromoacetyl-1,3-benzodioxolane, ethanone,1-1,3-benzodioxol-5-yl-2-bromo, 1-benzo d 1,3 dioxol-5-yl-2-bromoethanone, 1-benzo 1,3 dioxol-5-yl-2-bromo-ethanone, 1-2h-1,3-benzodioxol-5-yl-2-bromoethanone, ethanone, 1-1,3-benzodioxol-5-yl-2-bromo, 1-2h-benzo 3,4-d 1,3-dioxolen-5-yl-2-bromoethan-1-one
PubChem CID 243777
IUPAC Name 1-(1,3-benzodioxol-5-yl)-2-bromoethanone
SMILES C1OC2=C(O1)C=C(C=C2)C(=O)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 243.06
Percent Purity 97%
Chemical Name or Material 1-(1, 3-Benzodioxol-5-yl)-2-bromoethan-1-one