missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethylguanidine, 99%

Catalog No. AC221295000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221295000 500 g Glass bottle
AC221291000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC221295000 Supplier Thermo Scientific Chemicals Supplier No. 221295000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 80-70-6 | C5H13N3 | 115.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80-70-6
Molecular Weight (g/mol) 115.18
InChI Key KYVBNYUBXIEUFW-UHFFFAOYSA-N
Synonym tetramethylguanidine, n,n,n',n'-tetramethylguanidine, guanidine, n,n,n',n'-tetramethyl, tetramethyl guanidine, unii-vez101e7zu, 1,1,3,3-tetramethyl guanidine, n,n-1,1,3,3-tetramethylguanidine, vez101e7zu, 1,1,3,3-tetramethyl-guanidine, n,n,n',n'-tetramethyl-guanidine
PubChem CID 66460
SMILES CN(C)C(=N)N(C)C

Specifications

Melting Point -30.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 160.0°C to 162.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H13N3
Linear Formula (CH3)2NC(=NH)N(CH3)2
Refractive Index 1.4670 to 1.4690
Quantity 500 g
Beilstein 04,I,335
Fieser 01,1145; 04,489; 12,477
Merck Index 15,9375
Specific Gravity 0.91
Solubility Information Solubility in water: miscible. Other solubilities: miscible
IUPAC Name 1,1,3,3-tetramethylguanidine
Viscosity 0.002 Pas (20°C)
Formula Weight 115.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetramethylguanidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remov
EINECSNumber 201-302-7

RUO – Research Use Only