missing translation for 'onlineSavingsMsg'
Learn More

1-(1,2,3,6-Tetrahydro-4-pyridyl)-2-benzimidazolinone 97.0+%, TCI America™

Catalog No. T30851G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T30851G Supplier TCI America Supplier No. T30851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2147-83-3
Molecular Formula C12H14N3O
Molecular Weight (g/mol) 216.26
MDL Number MFCD00044827
InChI Key YFEOSTXFQCDCAR-UHFFFAOYSA-O
Synonym 1,3-dihydro-1-1,2,3,6-tetrahydro-4-pyridinyl-2h-benzimidazole-2-one, 1,3-dihydro-1-1,2,3,6-tetrahydro-4-pyridinyl-2h-benzimidazol-2-one, 3-1,2,3,6-tetrahydropyridin-4-yl-1h-benzimidazol-2-one, 1-1,2,3,6-tetrahydro-4-pyridyl-2-benzimidazolinone, 1-1,2,3,6-tetrahydropyridin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 1-1,2,3,6-tetrahydropyridin-4-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-4-1,2,5,6-tetrahydropyridyl-3-hydrobenzimidazol-2-one, 1-1,2,3,6-tetrahydropyridin-4-yl-3h-1,3-benzodiazol-2-one, benzoimidazol-2-one, 1-1,2,3,6-tetrahydro-pyridin-4-yl-1,3-dihydro
PubChem CID 75070
IUPAC Name 4-(2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)-1,2,3,6-tetrahydropyridin-1-ium
SMILES O=C1NC2=CC=CC=C2N1C1=CC[NH2+]CC1

Specifications

Melting Point 196°C
Color Yellow
Quantity 1 g
Formula Weight 215.26
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(1,2,3,6-Tetrahydro-4-pyridyl)-2-benzimidazolinone