missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrachloroethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D

Catalog No. AC464210075
Change view
Click to view available options
Quantity:
7.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464210075 7.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC464210075 Supplier Thermo Scientific Chemicals Supplier No. 464210075

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33685-54-0 | C2H2Cl4 | 169.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33685-54-0
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 169.85
InChI Key QPFMBZIOSGYJDE-QDNHWIQGSA-N
IUPAC Name tetrachloro(²H₂)ethane
SMILES [2H]C(Cl)(Cl)C([2H])(Cl)Cl

Specifications

Quantity 7.5 mL
Density 1.6100 g/mL
Refractive Index 1.4924
Specific Gravity 1.61
Boiling Point 146.0°C
Linear Formula (Cl2CD)2
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in ether, chloroform, benzene and alcohols
Formula Weight 169.86
Grade NMR
Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Fatal if inhaled.

Fatal in contact with skin.

Toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

Remove/Take off immediately all contaminated clothing.

IF ON SKIN: Gently wash with plenty of soap and water.

Immediately call a POISON CENTER or doctor/physician.

IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

Avoid release to the environment.

EINECSNumber 251-634-1

RUO – Research Use Only