missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide 98.0+%, TCI America™

Catalog No. H10565G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10565G 5 g
H10561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10565G Supplier TCI America Supplier No. H10565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84246-29-7
Molecular Formula C3HF6NO4S2
Molecular Weight (g/mol) 293.15
MDL Number MFCD06200806
InChI Key WOAGDWWRYOZHDS-UHFFFAOYSA-N
Synonym 4,4,5,5,6,6-Hexafluoroperhydro-1,3,2-dithiazine 1,1,3,3-Tetraoxide
PubChem CID 11208600
IUPAC Name 4,4,5,5,6,6-hexafluoro-1λ⁶,3λ⁶,2-dithiazinane-1,1,3,3-tetrone
SMILES FC1(F)C(F)(F)S(=O)(=O)NS(=O)(=O)C1(F)F

Specifications

Color White
Quantity 5 g
UN Number 1759
Formula Weight 293.15
Percent Purity ≥98.0% (T)
Physical Form Crystals
Chemical Name or Material 1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide