missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2,2,3,3-Hexafluoro-1-(trifluoromethoxy)-3-[(1,2,2-trifluorovinyl)oxy]propane 97.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 64°C |
| Quantity | 5 g |
| Formula Weight | 332.04 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,1,2,2,3,3-Hexafluoro-1-(trifluoromethoxy)-3-[(1,2,2-trifluorovinyl)oxy]propane |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |