missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1-Tris(4-hydroxyphenyl)ethane 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 248°C |
| Color | White |
| Quantity | 500 g |
| UN Number | 3077 |
| Formula Weight | 306.36 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,1,1-Tris(4-hydroxyphenyl)ethane |
Safety and Handling
| EINECSNumber | (4)-1668 |
| RTECSNumber | SL4375275 |
| TSCA | Yes |