missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-4-iodobutane 98.0+%, TCI America™

Catalog No. T25295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25295G 5 g
T252925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25295G Supplier TCI America Supplier No. T25295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 461-17-6
Molecular Formula C4H6F3I
Molecular Weight (g/mol) 237.992
MDL Number MFCD00041543
InChI Key LNDGACQEAYKNOI-UHFFFAOYSA-N
Synonym 1-iodo-4,4,4-trifluorobutane, 4,4,4-trifluoro-1-iodobutane, 4,4,4-trifluorobutyl iodide, 4-iodo-1,1,1-trifluorobutane, butane, 1,1,1-trifluoro-4-iodo, 1,1,1-trifluoro-4-iodo-butane, acmc-209k5v, 4,4,4-trifluorobutyliodide, ksc235m2j, 1-iodo-4,4,4-trifluoro-butane
PubChem CID 2775189
IUPAC Name 1,1,1-trifluoro-4-iodobutane
SMILES C(CC(F)(F)F)CI

Specifications

Boiling Point 128°C
Quantity 5 g
Formula Weight 237.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1-Trifluoro-4-iodobutane