missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-4-iodobutane, 98+%, stab. with copper

Catalog No. AAB2381106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2381106 5 g
AAB2381103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2381106 Supplier Thermo Scientific Chemicals Supplier No. B2381106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 461-17-6 | C4H6F3I | 237.992 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 461-17-6
Molecular Formula C4H6F3I
Molecular Weight (g/mol) 237.992
MDL Number MFCD00041543
InChI Key LNDGACQEAYKNOI-UHFFFAOYSA-N
Synonym 1-iodo-4,4,4-trifluorobutane, 4,4,4-trifluoro-1-iodobutane, 4,4,4-trifluorobutyl iodide, 4-iodo-1,1,1-trifluorobutane, butane, 1,1,1-trifluoro-4-iodo, 1,1,1-trifluoro-4-iodo-butane, acmc-209k5v, 4,4,4-trifluorobutyliodide, ksc235m2j, 1-iodo-4,4,4-trifluoro-butane
PubChem CID 2775189
IUPAC Name 1,1,1-trifluoro-4-iodobutane
SMILES C(CC(F)(F)F)CI

Specifications

Density 1.85
Boiling Point 126°C to 127°C
Flash Point >100°C (212°F)
Refractive Index 1.4335
Linear Formula CF3(CH2)3I
Quantity 5 g
Beilstein 1736064
Sensitivity Light sensitive
Formula Weight 237.99
Percent Purity ≥98%
Chemical Name or Material 1,1,1-Trifluoro-4-iodobutane, stabilized with copper
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only