missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,5,5,5-Hexafluoroacetylacetone, 99%

Catalog No. AC218531000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218531000 100 g Glass bottle
AC218530050 5 g Glass bottle
AC218530250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC218531000 Supplier Thermo Scientific Chemicals Supplier No. 218531000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1522-22-1 | C5H2F6O2 | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1522-22-1
Molecular Formula C5H2F6O2
Molecular Weight (g/mol) 208.06
InChI Key QAMFBRUWYYMMGJ-UHFFFAOYSA-N
Synonym hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hfac, 1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,3-bis trifluoromethyl propane-1,3-dione, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane
PubChem CID 73706
IUPAC Name 1,1,1,5,5,5-hexafluoropentane-2,4-dione
SMILES C(C(=O)C(F)(F)F)C(=O)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.4700g/mL
Boiling Point 70.0°C to 71.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.3320 to 1.3340
Linear Formula CF3COCH2COCF3
Quantity 100 g
Beilstein 01,III,3123
Specific Gravity 1.47
Solubility Information Solubility in water: immiscible
Formula Weight 208.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,1,1,5,5,5-Hexafluoroacetylacetone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 216-191-

RUO – Research Use Only