missing translation for 'onlineSavingsMsg'
Learn More

1,1'-[1,4-Phenylenebis(methylene)]bis(4,4'-bipyridinium) Dibromide 98.0+%, TCI America™

Catalog No. P14051G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14051G 1 g
P1405100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14051G Supplier TCI America Supplier No. P14051G

Chemical Identifiers

CAS 106867-97-4
Molecular Formula C28H24Br2N4
Molecular Weight (g/mol) 576.34
MDL Number MFCD03844804
InChI Key QSNYIUIZTNLTGX-UHFFFAOYSA-L
Synonym 1,1′C-(p-Xylylene)bis(4,4′C-bipyridinium) Dibromide
PubChem CID 44630455
IUPAC Name 1-{[4-({[4,4'-bipyridin]-1-ium-1-yl}methyl)phenyl]methyl}-[4,4'-bipyridin]-1-ium dibromide
SMILES [Br-].[Br-].C(C1=CC=C(C[N+]2=CC=C(C=C2)C2=CC=NC=C2)C=C1)[N+]1=CC=C(C=C1)C1=CC=NC=C1

Specifications

Quantity 1 g
Formula Weight 576.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-[1,4-Phenylenebis(methylene)]bis(4,4′-bipyridinium) Dibromide