missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexamethyldisilazane, HMDS, EMD-Millipore

Catalog No. M1121860100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1121860100 100 mL Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1121860100 Supplier Technidata Supplier No. 1.12186.0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 999-97-3
Molecular Formula C6H19NSi2
Molecular Weight (g/mol) 161.395
InChI Key FFUAGWLWBBFQJT-UHFFFAOYSA-N
Synonym Bis(trimethylsilyl)amine, HMDS
PubChem CID 13838
ChEBI CHEBI:85068
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane
SMILES C[Si](C)(C)N[Si](C)(C)C

Specifications

Packaging Glass
Explosion Limit 0.8 to 25.9 %(V)
Ignition Point 325°C
Vapor Pressure 20 hPa (20°C)
Viscosity 0.9mm2/sec., kinematic (25°C)
Density Density: 0.774g/cm3 (20°C)
Quantity 100 mL
Formula Weight 161.39
Chemical Name or Material 1,1,1,3,3,3-Hexamethyldisilazane
Recommended Storage Store at 15° to 25°C