missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroisopropyl methacrylate, 99%, stab.

Catalog No. AAB2144714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2144714 25 g
AAB2144706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2144714 Supplier Thermo Scientific Chemicals Supplier No. B2144714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 3063-94-3 | C7H6F6O2 | 236.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3063-94-3
Molecular Formula C7H6F6O2
Molecular Weight (g/mol) 236.113
MDL Number MFCD00040105
InChI Key FMQPBWHSNCRVQJ-UHFFFAOYSA-N
Synonym hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-hexafluoroisopropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, 2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate, hexafluoro-2-propyl methacrylate, 1h-1-trifluoromethyl trifluoroethyl methacrylate, 1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate, 1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate, hfipma
PubChem CID 76469
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F

Specifications

Density 1.302
Boiling Point 99°C
Flash Point 14°C (57°F)
Refractive Index 1.331
Quantity 25 g
UN Number UN1993
Formula Weight 236.12
Percent Purity 99%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl methacrylate, stabilized
Hazard Category H224-H315-H319-H335
Hazard Statement GHS H Statement
H224-H315-H319-H335
Extremely flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 221-309-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only