missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroisopropyl acrylate, 98+%, stab. with 50ppm 4-methoxyphenol

Catalog No. AAL1212814
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1212814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1212814 Supplier Thermo Scientific Chemicals Supplier No. L1212814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used in agrochemical, pharmaceutical and dyestuff field. | CAS: 2160-89-6 | C6H4F6O2 | 222.09 g/mol

1,1,1,3,3,3-Hexafluoroisopropyl acrylate is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2160-89-6
Molecular Formula C6H4F6O2
Molecular Weight (g/mol) 222.09
MDL Number MFCD00040104
InChI Key MNSWITGNWZSAMC-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoroisopropyl acrylate, hexafluoroisopropyl acrylate, 1,1,1,3,3,3-hexafluoropropan-2-yl acrylate, 2-propenoic acid, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, acrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, hexafluoro-2-propyl acrylate, mnswitgnwzsamc-uhfffaoysa, 1h-1-trifluoromethyl trifluoroethyl acrylate
PubChem CID 75096
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl prop-2-enoate
SMILES FC(F)(F)C(OC(=O)C=C)C(F)(F)F

Specifications

Color Colorless
Density 1.37
Boiling Point 81°C to 82°C
Flash Point 17°C (62°F)
Packaging Glass bottle
Refractive Index 1.319
Linear Formula H2C=CHCO2CH(CF3)2
Quantity 25 g
UN Number UN3272
Beilstein 2094742
Specific Gravity 1.37
Solubility Information Not miscible in water.
Formula Weight 222.09
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl acrylate
Show More Show Less
Hazard Category H225-H315-H319-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 218-479-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only