missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol 99.0+%, TCI America™

Catalog No. H07245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07245G 5 g
H072425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07245G Supplier TCI America Supplier No. H07245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 718-64-9
Molecular Formula C9H6F6O
Molecular Weight (g/mol) 244.136
MDL Number MFCD00040983
InChI Key IZPIZCAYJQCTNG-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-phenyl-2-propanol, hexafluoro-2-hydroxyisopropyl benzene, bis trifluoromethyl phenylcarbinol, 2,2,2,2',2',2'-hexafluorocumyl alcohol, alpha,alpha-bis trifluoromethyl benzyl alcohol, hexafluoro-2-phenylisopropanol, benzenemethanol, .alpha.,.alpha.-bis trifluoromethyl, hexafluoro-2-phenyl isopropanol, hexafluoro-2-phenyl-2-propanol, benzenemethanol, alpha,alpha-bis trifluoromethyl
PubChem CID 69748
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-ol
SMILES C1=CC=C(C=C1)C(C(F)(F)F)(C(F)(F)F)O

Specifications

Color Colorless
Boiling Point 160°C
Quantity 5 g
UN Number 1987
Formula Weight 244.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol