missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 0129 Discs

Catalog No. OXDD0015T
Change view
Click to view available options
Concentration:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
OXDD0015T 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXDD0015T Supplier Thermo Scientific™ Supplier No. DD0015T

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Differentiate Vibrios from other Gram-negative rods using these diagnostic discs.

Differentiate vibrios from other Gram-negative rods using Thermo Scientific™ Oxoid™ 0129 Discs. The test exploits the sensitivity of vibrios to the vibrio static agent 0129 (2,4-Diamino-6,7-di-iso-propylpteridine phosphate)1. 0129 Discs are of great value in differentiating vibrios from other Gram-negative rods and particularly from aeromonads, which are characteristically resistant to 0129. The degree of sensitivity of vibrios to 0129 can also be used as a diagnostic feature in differentiation of Vibrio species.

50 Discs/Cartridge.

Specifications

Content And Storage 2°C to 8°C
Concentration 150 μg
Detectable Analytes Vibrio species, Gram-negative rods
Format Disk, Cartridge
Type O129 Disc
Packaging Type 50 Discs/Cartridge
Quantity 150μg