Learn More
Laxco 0.5X Supplementary Objective for Z200PRO Series Stereo Microscope
Description
- 0.5X stereo microscope assisted objective lens, effectively condenses the field of view
- Halves the magnification of original objectives, a very practical objective lens accessory to have
- Easy installation on the microscope, just screw it on
- Significantly increase working distance up to 165 mm enabling user to view larger samples
Spécifications
Spécifications
| Type | Auxiliary Lens |
| Autoclavable | Not Autoclavable |
| Sterility | Sterile |