missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 0.5 mL Conical Screw Cap Tubes: Presterilized
Features chemical resistant O-ring
Supplier: Axygen™ SCT050RS
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
- Presterilized cap tubes have non-looped caps attached with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
- Available in range of colors for easy sample identification
Specifications
| RNase, DNase, pyrogen-free | |
| Polypropylene | |
| Polypropylene | |
| O-ring | |
| Sterile | |
| Conical |
| Red | |
| 0.5 mL | |
| Non-looped | |
| 500 Tubes and caps/Pk.; 8 Packs/Cs. | |
| Screw Cap | |
| 20,000 x g |