missing translation for 'onlineSavingsMsg'
Learn More

0.1% Formic Acid in Acetonitrile, Solstice

Catalog No. 60090010
Change view
Click to view available options
4X4L GLASS BOTTLE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
60090010 4X4L GLASS BOTTLE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60090010 Supplier Honeywell Inc Supplier No. LC4414X4L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

LabReady™ Solvent Blend, for LC-MS and HPLC

LabReady™ Solvent Blends

  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Solstice Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Solstice LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Solstice's high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Specifications

CAS 64-18-6, 75-05-8
CAS Max % 0.10%
CAS Min % >99.00%
Quantity 4 x 4 L
Flash Point 8°C
Packaging Glass Bottle
Physical Form Liquid
Purity Grade Notes LC-MS grade Labready™™ blends
Boiling Point 82°C