Thermo Scientific Water Analysis Solutions
Thermo Scientific Orion Versa Star Pro Multiparameter Benchtop Meter
Meet your most challenging applications for pH, ion concentration, conductivity, dissolved oxygen, and temperature with the accuracy and flexibility of the Thermo Scientific™ Orion™ Versa Star Pro™ Benchtop Meter.
- Customize the four meter channels with interchangeable modules
- View measurements on the color display
- Log up to 2,000 data point sets with time and date stamps
- Print or download data via USB or RS232
Modules
- pH/ISE
- Conductivity
- Dissolved oxygen/RDO
Thermo Scientific Benchtop Meters
pH Calibration & Testing with the Thermo Scientific Orion Versa Star
Gayle Gleichauf, Applications Lab Manager, calibrates and tests the pH of local drinking water, using the Thermo Scientific Orion Versa Star benchtop meter. The meter can be set up to include the electrode serial number, the calibration with three buffers, and incremental sample number (pH, millivolts, temperature, method, and more).
Fluoride Calibration & Testing with the Thermo Scientific Orion Versa Star
Gayle Gleichauf, Applications Lab Manager, tests for fluoride in the local tap water, using the Thermo Scientific Orion Versa Star benchtop meter. She demonstrates the various functionalities of the meter, including setting customization, calibration dates, the electrode condition, and more.
| Meter | Display Type | Keypad | Number of Channels | Meter-powered Stirrer Probe Inputs | Datalog | USB Output | RS232 Output | |
|---|---|---|---|---|---|---|---|---|
Customizable color graphic LCD with step-by-step text prompts | Numeric with menu-specific function keys | 1 to 4 | 2 | 2000 with date/time | 2 | 1 | Buy Now | |
Graphic LCD with step-by-step text prompts | Comprehensive with menu-specific function keys | 1 or 2 | 1 | 2000 with date/time | 1 | 1 | Buy Now | |
Graphic LCD with step-by-step text prompts | Numeric with menu-specific function keys | 2 | 1 | 1000 with date/time | 1 | 1 | Buy Now | |
Fixed segment LCD with basic text prompts | Simple with highlighted measure key | 1 | 50 | N/A | N/A | N/A | Buy Now |
Thermo Scientific Portable Meters
| Meter | Display Type | Keypad | Number of Channels | Datalog | USB Output | RS232 Output | Date/Time, GLP | - |
|---|---|---|---|---|---|---|---|---|
Graphic LCD with backlight | Comprehensive with menu-specific function keys | 1 to 4 | 5000 with date/time | 1 | 1 | Yes | Buy Now | |
Graphic LCD with backlight | Comprehensive with menu-specific function keys | 1 | 1000 with date/time | 1 | 1 | Yes | Buy Now | |
Fixed-segment LCD | Simple with highlighted measure key | 1 | 50 | N/A | N/A | N/A | Buy Now |
Thermo Scientific Orion Electrodes
The Thermo Scientific™ Orion™ electrode line offers a wide variety of pH, conductivity, ORP and ISE electrodes to deliver accurate, quick and reproducible measurements.
For pH measurement, Ross combination pH electrodes offer unsurpassed accuracy and precision and are compatible with any brand of meter.
To ensure you choose the electrode that will suit your needs, you will need to consider body materials, body styles and sample requirements. Use this interactive selection tool to help you choose the correct electrode for your application.
Epoxy or Glass?
Epoxy Electrodes
- Extremely durable, rugged body minimizes breakage
- Electrode bulb protected by bulb guard
- Low-maintenance, gel-filled; can be used in higher temperatures than glass electrodes
Glass Electrodes
- Compatible with virtually any sample, including solvents
- Easy to clean
- Can handle higher temperatures
Water Analysis Solutions
Water Analysis Solutions Demonstration Videos
pH Calibration & Testing with the Thermo Scientific Orion Versa Star
Gayle Gleichauf, Applications Lab Manager, calibrates and tests the pH of local drinking water, using the Thermo Scientific Orion Versa Star benchtop meter. The meter can be set up to include the electrode serial number, the calibration with three buffers, and incremental sample number (pH, millivolts, temperature, method, and more).
Fluoride Calibration & Testing with the Thermo Scientific Orion Versa Star
Gayle Gleichauf, Applications Lab Manager, tests for fluoride in the local tap water, using the Thermo Scientific Orion Versa Star benchtop meter. She demonstrates the various functionalities of the meter, including setting customization, calibration dates, the electrode condition, and more.
Water Analysis Colorimetry
Orion AquaMate Spectrophotometers
Depending on the needs of your lab, choose the AquaMate 7000 Vis Spectrophotometer for 325 to 1100 nm wavelengths or choose the AquaMate 8000 UV-Vis with full 190 to 1100 nm wavelengths for a greater range of measurement options. Use your current chemistry reagents — no need to change. The AquaMate preprogrammed methods are compatible with many leading reagent manufacturers.
Orion AquaMate Spectrophotometers
| Spectrophotometers | Optical Design | Light Source (Typical lifetime) | Detector | Wavelength | Range | Accuracy | Repeatability | - |
|---|---|---|---|---|---|---|---|---|
Dual beam — internal reference detector | 1.8nm | Xenon flash lamp (5 years) | Dual silicon photodiodes | 190 to 1100nm | ±1.0nm | ±0.5nm | Buy Now | |
Single beam | 5.0nm | Tungsten-halogen lamp (1000 hours) | Silicon photodiode | 325 to 1100 nm | ±1.0nm | ±0.5nm | Buy Now |
Colorimeters
Thermo Scientific Orion Colorimeters provide portable design with uncompromised performance. Thermo Scientific Orion meters are accurate, dependable and easy to use, with handheld portability, preprogrammed methods, optimum wavelengths, user-friendly displays and easy calibration.
Colorimeters
| Instrument | Wavelengths | Sample Cell Shape (Sizes) | Test Reagent Formats | Buy Now |
|---|---|---|---|---|
430, 530, 560, 580, 610, and 660nm | Round (13, 16, and 24mm) | Powder, tablet, liquid, reagent tubes, digestion tubes; no reagents required for color, turbidity, and suspended solids tests | Buy Now | |
420, 520, 580, and 610nm | Round (13, 16, and 24mm) | Buy Now | ||
530nm | Round (10 and 16mm) | Powder packs, bulk powder dispensers | Buy Now | |
430nm and 610nm | Round (13, 16, and 20mm) | Digestion tubes | Buy Now |
Standards
| Standard | Description | uy Now |
|---|---|---|
AQUAfast 1,000 ppm COD standard, 475mL | Buy Now | |
AQUAfast 10,000 ppm COD standard, 475mL | Buy Now |
Turbidity Meters
Thermo Scientific Orion offers turbidity meters. Choose from the Orion AQ4500 Turbidity Meter which offers an all-in-one capability, featuring both white and infrared light sources. The Orion AQ3010 Turbidity Meter, includes a field case, offers faster calibration with step-by-step prompts.
Turbidity Meters
| Meters | Incident Light Source | Measurement Modes | Measurement Ranges | Resolution | Sample Cell Compatibility | PowerHeader | Warranty | Buy Now |
|---|---|---|---|---|---|---|---|---|
Dual Source (White light LED and IR LED) | EPA 180.1, ISO nephelometric, ISO absorbance, IR ratiometric, % transmittance, EPA GLI method 2, EBC (European Brewing Chemists), ASBC (American Society of Brewing Chemists) | 0 to 4000 NTU EPA 0 to 2000 NTU GLI range 0 to 40 NTU ISO-7027 0 to 150 FAU IR Ratio 0 to 4000 NTU EBC 0 to 24.5 ASBC 0 to 236 | 0.01 (0 to 9.99 NTU), 0.1 (10 to 99.9), 1 (100 to 4000) | 24mm round | 4 AA batteries | 2 years | Buy Now | |
IR LED | ISO nephelometric | 0.01 to 1000 NTU | 0.01 (0.01 to 19.99 NTU), 0.1 (20 to 99.9), 1 (100 to 1000) | 25mm round | 4 AAA batteries | 3 years | Buy Now |
Fisher Scientific Results Wastewater Workflow Guide
View the workflow for equipment that is essential to monitor and control water treatment processes in industrial and wastewater treatment plants.
Fisher Scientific Results Potable Water Workflow Guide
View the workflow for equipment that is essential to monitor and control the water treatment process in potable water treatment plants.
pH Measurement per USP 791 Webinar
A webinar highlighting the updated USP 791 standard test method and its requirements for the pH measurement system.