CAS RN 371924-24-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ MCU i4
Negative modulator of mitochondrial Ca2+ uniporter (MCU)