missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MCU i4
Description
MCU i4 is a negative modulator of mitochondrial calcium (Ca2+) uniporter (MCU) complex, which binds to MICU1, a regulatory protein within MCU complex. MCU i4 reduces mitochondrial Ca2+ uptake in multiple cell lines including MEFs, MDA-MB,231 and HEK293T cells, and demonstrates a negative effect on mitochondrial membrane potential. In ex vivo muscle fibres, MCU i4 reduces mitochondrial Ca2+ uptake and impairs muscle cell growth.
Specifications
Specifications
| Chemical Name or Material | 4-[[4-(Diethylamino)phenyl]amino]-6-methyl-3-quinolinecarboxylic acid ethyl ester |
| CAS | 371924-24-2 |
| Quantity | 50 mg |
| Target | Mitochondrial Calcium Uniporter Inhibitors |
| Molecular Formula | C23H27N3O2 |
| Purity | 98% |