Get a Pipette with Eppendorf Microcentrifuge Purchase
New Lab Start-up Program
When you buy a qualifying ventilated Eppendorf™ 5430 Microcentrifuge equipped with a fixed-angle 30 x 2 mL rotor (Cat. No. 14-280-104), you’ll get a redeemable Eppendorf™ Repeater™ E3 Pipette and an assortment pack of Eppendorf™ Combitips™ at no additional cost. To take advantage of this offer, order using the catalog numbers listed below, log in to the Redemption Center, and complete the form to receive your products.
Features and Benefits
The compact ventilated Eppendorf 5430 Centrifuge offers the versatility of a multipurpose centrifuge in a small footprint. It automatically recognizes up to 12 compatible rotors for processing microtubes, PCR strips, microplates, and larger conical tubes. The Eppendorf Repeater E3 Pipette reduces fatigue by decreasing user movement and dispensing up to 100 times from a fully filled tip.
Get a Pipette with Eppendorf Microcentrifuge Purchase
Redeem Now
Promotion: 287
You must apply and qualify for NLSU to redeem this promotion.
Qualifying Products
    Catalog Number Description Price Quantity & Availability  
Success
05-403-94 Details
Eppendorf™ Centrifuge 5430 - Microcentrifuge Includes: 30 x 2.0 mL Aerosol-tight Rotor
Product Specifications
Width (Metric) Exterior 33 cm
Length (Metric) Exterior 42 cm
Run Time 30 sec. to 99 hr. 59 min. with HOLD
Maximum Speed Range 17,500 rpm
Dimensions (HWD) 10 x 13.2 x 16.8 in. (25 x 33 x 42 cm)
Dimensions (L x W x H) Exterior 34 x 42 x 25 cm (11.2 x 16.3 x 9.8 in.)
Height (Metric) Exterior 25 cm
Product Line Eppendorf 5430
Includes 30 x 2.0 mL Aerosol-tight Rotor
Max. RCF 20,817 x g (w/ incl. rotor)
Voltage 120 V
Width (English) Exterior 11.2 in.
Rotor Included Yes
Controller Type Keypad
Maximum Force Range 30,130 x g
Refrigerated No
Height (English) Exterior 9.8 in
Display Large, Brightly Lit LCD
Rotor FA-45-30-11
Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
Timer 30 sec. to 99:59 hr., Continuous Run Function
Ventilated Yes
Profile (Acceleration/Braking) Variable speed control from 100 to 17,500 rpm in 100 rpm increments
Net Weight 63.8 lb. (29 kg)
Electrical Requirements 120 V, 50/60 Hz (US)
Program Storage 50-program memory with 5 keys for one-touch recall of routine programs
Max. Speed 14,000 rpm (w/ incl. rotor)
Length (English) Exterior 16.8 in.
Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition
Standards EN ISO 12100, EN 50581, EN 61010-1, IEC 61010-2-020, EN 61326-1, EN 55011, EN 378-1, EN 378-2, 47 CFR FCC part 15, EN 50581, UL 61010-1, CAN/CSA-C22.2 No. 61010-1 (refer to manual for more details)
Capacity 30 x 1.5/2.0 mL Tubes (w/ incl. rotor)
Wattage 475 W
13-690-000 Details
Eppendorf™ Centrifuge 5430 - Microcentrifuge Includes: 30 x 2.0 mL Aerosol-tight Rotor
Product Specifications
Width (Metric) Exterior 33 cm
Length (Metric) Exterior 42 cm
Run Time 30 sec. to 99 hr. 59 min. with HOLD
Maximum Speed Range 17,500 rpm
Dimensions (HWD) 10 x 13.2 x 16.8 in. (25 x 33 x 42 cm)
Dimensions (L x W x H) Exterior 34 x 42 x 25 cm (11.2 x 16.3 x 9.8 in.)
Height (Metric) Exterior 25 cm
Product Line Eppendorf 5430
Includes 30 x 2.0 mL Aerosol-tight Rotor
Max. RCF 20,817 x g (w/ incl. rotor)
Voltage 120 V
Width (English) Exterior 11.2 in.
Rotor Included Yes
Controller Type Knob
Maximum Force Range 30,130 x g
Refrigerated No
Height (English) Exterior 9.8 in
Display Large, Brightly Lit LCD
Rotor FA-45-30-11
Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
Timer 30 sec. to 99:59 hr., Continuous Run Function
Ventilated Yes
Profile (Acceleration/Braking) Variable speed control from 100 to 17,500 rpm in 100 rpm increments
Net Weight 63.8 lb. (29 kg)
Electrical Requirements 120 V, 50/60 Hz (US)
Program Storage 50-program memory with 5 keys for one-touch recall of routine programs
Max. Speed 14,000 rpm (w/ incl. rotor)
Length (English) Exterior 16.8 in.
Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition
Standards EN ISO 12100, EN 50581, EN 61010-1, IEC 61010-2-020, EN 61326-1, EN 55011, EN 378-1, EN 378-2, 47 CFR FCC part 15, EN 50581, UL 61010-1, CAN/CSA-C22.2 No. 61010-1 (refer to manual for more details)
Capacity 30 x 1.5/2.0 mL Tubes (w/ incl. rotor)
Wattage 475 W
Redeemable Product
Catalog NumberDescription
Catalog Number
Description
Eppendorf™ Repeater™ E3 Pipette, 1 to 50 mL