UserName
Request Information About Labconco Fume Hoods
Request Information About Labconco Fume Hoods
Choose the Right Labconco Fume Hood for Your Lab
Labconco™ Protector™ XL Fume Hoods offer superior containment and come in a variety of sizes to fit any lab. They are currently in stock for fast shipping from a Fisher Scientific warehouse.
Designed for general chemistry applications, Protector XL Fume Hoods exceed the SEFA 1 standard for low-velocity high-performance hoods and can operate at face velocities as low as 60fpm. They offer the lowest air volume requirements on the market.
Protector XL Hoods have chemical-resistant and fiberglass-reinforced panel liners and baffles, safety-glass sashes, and powder-coated steel exteriors. The baffles can be removed for cleaning and the panels allow easy access to electrical and plumbing connections.
Customize your Protector XL Hood with additional fixture kits. Standard hoods can accommodate up to eight service fixtures and four electrical duplexes.
Catalog NumberDescription
16-317-155
Protector XL Benchtop Hood, Exterior Dimensions: 4 ft. x 31.7 in., with 2 Service Fixtures and 1 GFCI Duplex, 110V/60Hz
16-317-156
Protector XL Benchtop Hood, Exterior Dimensions: 6 ft. x 31.7 in., with 2 Service Fixtures and 1 GFCI Duplex, 110V/60Hz
16-317-157
Protector XL Benchtop Hood, Exterior Dimensions: 8 ft. x 31.7 in., with 2 Service Fixtures and 2 GFCI Duplexes, 110V/60Hz
Contact Us
Fill out the form below if you’d like a sales representative to contact you about Labconco Fume Hoods.
Required Field *
Company
Address