Gibco Peptone Starter Paks
Enhance your bioproduction applications with the proven history of consistent, reliable, high-performing Gibco™ peptones. Choose from the Gibco™ Starter Paks below for conveniently packaged samples of the peptones most commonly used in mammalian cell culture and microbial fermentation bioproduction processes. They’ll help you quickly identify the correct peptones, optimize your process, and achieve higher cell titers and yield.
Gibco™ Starter Pak No. 1
Peptones for Human and Animal Health Applications
A combination of animal origin-free yeast and soy-based peptones with an established history of use in vaccines, monoclonal antibodies, and recombinant protein production.
The kit includes five 100g samples*:
- Gibco™ Difco™ TC Yeastolate™ UF (Cat. No. B92804)
- Gibco™ Bacto™ TC Yeastolate™ (Cat. No. DF5577-15-5)
- Gibco™ Difco™ Yeast Extract UF(Cat. No. DF210929)
- Gibco™ Bacto™ Yeast Extract, Technical (Cat. No. DF0886-17-0)
- Gibco™ Difco™ Phytone™ Supplement UF (Cat. No. DF210931)
Gibco™ Starter Pak No. 2
Peptones for Vaccine Production
Animal origin and animal origin-free peptones offering many of the essential nutrients needed to produce human and animal vaccines.
The kit includes six 100g samples*:
- Gibco™ Bacto™ Yeast Extract (Cat. No. DF0127-17-9)
- Gibco™ Phytone™ Peptone (Cat. No. B11906)
- Gibco™ Difco™ Soytone(Cat. No. 21-248-8)
- Gibco™ Bacto™ Proteose™ Peptone No. 2 (Cat. No. DF0121-17-5)
- Gibco™ Bacto™ Proteose™ Peptone No. 3 (Cat. No. DF0122-17-4)
- Gibco™ Bacto™ Casamino Acids (Cat. No. DF0230-17-3)
Gibco™ Starter Pak No. 3
Animal Origin-Free Peptones for Vaccine Production
These products are suited for human and animal vaccine production and ideal for processes in which an animal origin-free medium is preferred.
The kit includes six 100g samples*:
- Gibco™ Bacto™ Yeast Extract(Cat. No. DF0127-17-9)
- Gibco™ Yeast Extract(Cat. No. B11929)
- Gibco™ Bacto™ TC Yeastolate (Cat. No. DF5577-15-5)
- Gibco™ Phytone™ Peptone (Cat. No. B11906)
- Gibco™ Difco™ Soytone (Cat. No. 21-248-8)
- Gibco™ Bacto Malt Extract (Cat. No. DF0186-17-7)
*Gibco Starter Paks are non-GMP and for evaluation use only. Referenced Cat. No. corresponds to additional GMP package sizes.