Intermountain Life Sciences: High-Purity Water for Pharmaceutical Purposes and GMP Solutions for Manufacturing
Water is widely used as a raw material, ingredient, and solvent in the processing, formulation, and production of pharmaceutical products, active pharmaceutical ingredients (APIs) and intermediates, compendial articles, and analytical reagents.
To address your need for cGMP-compliant solutions, we’ve expanded our state-of-the-art cGMP and ISO 13485:2016 certified facility in Salt Lake City, Utah, to meet or exceed regulatory and customer expectations.
The heart of our operation is a nine-step water-for-injection (WFI) purification system. Capable of producing more than 20,000 liters per day, we offer United States Pharmacopoeia (USP) water and saline for manufacturing and laboratory applications.
We also offer EP, cell culture, and molecular biology certified water, as well as buffered salt solutions. We use technology, business leadership, and logistics to make sure that you receive products that meet your specifications, delivery, and cost concerns.
Custom Manufacturing
We can manufacture custom formulations in a wide variety of packaging and delivery options for easier integration into today’s diverse manufacturing platforms.
- Upstream expression systems: solutions for mammalian, yeast, insect, and bacterial production platforms
- Downstream purification processes: solutions for protein, antibody, and cell/gene therapy workflows
- Cell and tissue processing solutions
- Molecular biology and diagnostic reagents
- Medical device manufacturing/processing solutions
Popular Products
USP Water for Injection (WFI)
USP grade terminally distilled water for injection is a sterile, non-pyrogenic preparation that contains no bacteriostat, antimicrobial agents, or added buffer. It complies with both USP monographs for water for injection and can be packaged in bulk for other commercial uses. Available in sizes from 500mL to 200L.
USP Water for Irrigation
USP grade water for irrigation is a sterile, hypotonic, and non-pyrogenic. It is prepared by distillation and contains no antimicrobial or bacteriostatic agents or added buffers. The pH is 5.7 (5.0 to 7.0), and the product complies with the USP monograph for water for irrigation. Available in sizes from 500mL to 200L.
Normal Saline, 0.9%
Intermountain Life Sciences 0.9% Sodium Chloride is a sterile aqueous solution for injection that contains no antimicrobial agents. It contains not less than 95% and not more than 105% of the labeled amount of sodium chloride (NaCl).
Content provided by: