accessibility menu, dialog, popup

UserName

CH3PCH3CH3

CAS RN 998-40-3

IUPAC Name: tributylphosphane
Synonyms: tributylphosphine
Molecular Weight (g/mol): 202.32
Molecular Formula: C12H27P
InChi Key: TUQOTMZNTHZOKS-UHFFFAOYSA-N
SMILES: CCCCP(CCCC)CCCC
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)