accessibility menu, dialog, popup

UserName

CAS RN 99306-64-6

CH3CH3H3COONHNH2NNH2OHNNOOHCl

CAS RN 99306-64-6

IUPAC Name: tert-butyl N-{4-[(diaminomethylidene)amino]-1-[(4-nitrophenyl)carbamoyl]butyl}carbamate hydrochloride
Synonyms: tert-butyl N-{4-[(diaminomethylidene)amino]-1-[(4-nitrophenyl)carbamoyl]butyl}carbamate hydrochloride
Molecular Weight (g/mol): 430.89
Molecular Formula: C17H27ClN6O5
InChi Key: OUVONNGVXPOXGB-UHFFFAOYNA-N
SMILES: Cl.CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results